Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Antibody detects endogenous levels of total IFN21.
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IFN-alpha-21; Interferon alpha-21; Interferon alpha-F; interferon, alpha 21; LeIF F; leukocyte interferon protein
Gene Aliases: IFN-alphaI; IFNA21; LeIF F; leIF-F
UniProt ID: (Human) P01568
Entrez Gene ID: (Human) 3452
Molecular Function: interferon superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support