Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 4
Antibody detects endogenous levels of total IFNA4/10/14.
IFNA4 (Interferon Alpha 4) and IFNA14 (Interferon Alpha 14) are predicted to enable cytokine activity and type I interferon receptor binding activity and to be involved in several processes, including B cell activation; lymphocyte activation involved in immune response; and positive regulation of peptidyl-serine phosphorylation of STAT protein. IFNA10 (Interferon Alpha 10) encodes a protein that belongs to the type I interferon family of proteins, and is located in a cluster of alpha interferon genes on chromosome 9. Interferons are small regulatory molecules that function in cell signaling in response to viruses and other pathogens or tumor cells. This gene is intronless and the encoded protein is secreted.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IFN-alpha-10; IFN-alpha-14; IFN-alpha-4; interferon alpha 14; interferon alpha family, gene 4; Interferon alpha-10; Interferon alpha-14; Interferon alpha-4; Interferon alpha-4B; Interferon alpha-6L; Interferon alpha-76; Interferon alpha-C; Interferon alpha-H; Interferon alpha-M1; Interferon lambda-2-H; interferon, alpha 10; interferon, alpha 14; interferon, alpha 4; LeIF C; LeIF H; MuIFN-alpha 4
Gene Aliases: Ifa4; IFN-alpha4a; IFN-alphaC; IFN-alphaH; IFNA10; IFNA14; IFNA4; INFA4; LEIF2H; OTTMUSG00000007662
UniProt ID: (Human) P01566, (Human) Q7M4S1, (Human) P05014, (Mouse) P07351
Entrez Gene ID: (Human) 3446, (Human) 3448, (Human) 3441, (Mouse) 404549, (Mouse) 15967
Molecular Function:
interferon superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support