Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IFN-alpha-4; interferon alpha family, gene 4; Interferon alpha-4; Interferon alpha-4B; Interferon alpha-76; Interferon alpha-M1; interferon, alpha 4; MuIFN-alpha 4
Gene Aliases: Ifa4; IFN-alpha4a; IFNA4; INFA4
UniProt ID: (Human) P05014, (Mouse) P07351
Entrez Gene ID: (Human) 3441, (Mouse) 15967
Molecular Function:
interferon superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support