Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
Interleukin 7 (IL-7) is a hematopoietic cytokine that is constitutively produced by stromal cells from the bone marrow and thymus. IL-7 signals through the IL-7R to promote the differentiation of hematopoietic stem cells to lymphoid precursor cells, which give rise to T cells, B cells, and natural killer cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H-IL-7; IL-7; ILN; Interleukin; Interleukin-7; Interleukin7; M-IL-7
Gene Aliases: IL-7; IL7
UniProt ID: (Human) P13232
Entrez Gene ID: (Human) 3574
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support