Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
Immunogen sequence: MAASGSGMAQ KTWELANNMQ EAQSIDEIYK YDKKQQQEIL AAKPWTKDHH YFKYCKISAL ALLKMVMHAR SGGNLEVMGL MLGKVDGETM IIMDSFALPV
JAB-1 is a 38 kDa protein which was originally described as a coactivator for cJUN. It is present in a large protein complex called the "Signalosome", which has been proposed to play a signaling role in mammalian cells. This protein complex appears to be analogous to the COP9 complex in plants, which is important in light-mediated plant development. Although the specific function of JAB-1 in this complex is unclear, it has recently been implicated in targeting proteins for degradation by the proteasome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 38 kDa Mov34 homolog; COP9 (constitutive photomorphogenic) homolog, subunit 5; COP9 complex S5; COP9 constitutive photomorphogenic homolog subunit 5; COP9 signalosome complex subunit 5; Jun activation domain-binding protein 1; Jun coactivator; Kip1 C-terminus-interacting protein 2; MGC3149; SGN5; signalosome subunit 5; testis secretory sperm-binding protein Li 231m
Gene Aliases: AI303502; COPS5; CSN5; JAB1; Kic2; MOV-34; Mov34; SGN5
UniProt ID: (Human) Q92905, (Mouse) O35864
Entrez Gene ID: (Human) 10987, (Rat) 312916, (Mouse) 26754
Molecular Function:
translation initiation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support