Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MRSPRTRGRS GRPLSLLLAL LCALRAKVCG ASGQFELEIL SMQNVNGELQ NGNCCGGARN PGDRKCTRDE CDTYFKVCLK EYQSRVTAGG PCSFGSGSTP VIGGNTFNLK ASRGNDRNRI VLPFSFAWPR SYTLLVEAWD SSNDTVQPDS IIEKASHSGM INPSRQWQTL KQNTGVAHFE YQIRVTCDDY YYGFGCNKFC RPRDDFFGHY ACDQNGNKTC MEGWMGPECN RAICRQGCSP KHGSCKLPGD CRCQYGWQGL YCDKCIPHPG CVHGICNEPW QCLCETNWGG QLCDKDLNYC GTHQPCLNGG TCSNTGPDKY QCSCPEGYSG PNCEIAEHAC LSDPCHNRGS CKETSLGFEC ECSPGWTGPT CSTNIDDCSP NNCSHGGTCQ DLVNGFKCVC PPQWTGKTCQ LDANECEAKP CVNAKSCKNL IASYYCDCLP GWMGQNCDIN INDCLGQCQN DASCRDLVNG YRCICPPGYA GDHCERDIDE CASNPCLNGG HCQNEINRFQ CLCPTGFSGN LCQLDIDYCE PNPCQNGAQC YNRASDYFCK CPEDYEGKNC SHLKDHCRTT PCEVIDSCTV AMASNDTPEG VRYISSNVCG PHGKCKSQSG GKFTCDCNKG FTGTYCHENI NDCESNPCRN GGTCIDGVNS YKCICSDGWE GAYCETNIND CSQNPCHNGG TCRDLVNDFY CDCKNGWKGK TCHSRDSQCD EATCNNGGTC YDEGDAFKCM CPGGWEGTTC NIARNSSCLP NPCHNGGTCV VNGESFTCVC KEGWEGPICA QNTNDCSPHP CYNSGTCVDG DNWYRCECAP GFAGPDCRIN INECQSSPCA FGATCVDEIN GYRCVCPPGH SGAKCQEVSG RPCITMGSVI PDGAKWDDDC NTCQCLNGRI ACSKVWCGPR PCLLHKGHSE CPSGQSCIPI LDDQCFVHPC TGVGECRSSS LQPVKTKCTS DSYYQDNCAN ITFTFNKEMM SPGLTTEHIC SELRNLNILK NVSAEYSIYI ACEPSPSANN EIHVAISAED IRDDGNPIKE ITDKIIDLVS KRDGNSSLIA AVAEVRVQRR PLKNRTDFLV PLLSSVLTVA WICCLVTAFY WCLRKRRKPG SHTHSASEDN TTNNVREQLN QIKNPIEKHG ANTVPIKDYE NKNSKMSKIR THNSEVEEDD MDKHQQKARF AKQPAYTLVD REEKPPNGTP TKHPNWTNKQ DNRDLESAQS LNRMEYIV
The jagged 1 protein encoded by JAG1 is the human homolog of the Drosophilia jagged protein. Human jagged 1 is the ligand for the receptor notch 1, the latter a human homolog of the Drosophilia jagged receptor notch. Mutations that alter the jagged 1 protein cause Alagille syndrome. Jagged 1 signalling through notch 1 has also been shown to play a role in hematopoiesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD339; jagged canonical Notch ligand 1; Jagged1; Protein jagged-1
Gene Aliases: AGS; AHD; AWS; CD339; HJ1; JAG1; JAGL1
UniProt ID: (Human) P78504
Entrez Gene ID: (Human) 182
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support