Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EELEPKRVRS SGPEAEEVQE EEELEEETGG EGPPAPIPTT GSPSTQENQN GLEPAEGTTS GPSAPFSTLT PRLH
Serine/threonine kinase which acts as a negative regulator of Ras-related Rap2-mediated signal transduction to control neuronal structure and AMPA receptor trafficking. Required for normal synaptic density, dendrite complexity, as well as surface AMPA receptor expression in hippocampal neurons. Can activate the JNK and MAPK14/p38 pathways and mediates stimulation of the stress-activated protein kinase MAPK14/p38 MAPK downstream of the Raf/ERK pathway. Phosphorylates: TANC1 upon stimulation by RAP2A, MBP and SMAD1. Has an essential function in negative selection of thymocytes, perhaps by coupling NCK1 to activation of JNK1.; Isoform 4 can activate the JNK pathway. Involved in the regulation of actin cytoskeleton reorganization, cell-matrix adhesion, cell-cell adhesion and cell migration.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [histone H3]-trimethyl-L-lysine(4) demethylase 5C; Histone demethylase JARID1C; JmjC domain-containing protein SMCX; Jumonji, AT rich interactive domain 1C (RBP2-like); Jumonji/ARID domain-containing protein 1C; lysine (K)-specific demethylase 5C; Lysine-specific demethylase 5C; Protein SmcX; Protein Xe169; sele; Smcx homolog, X chromosome; Smcy homolog, X-linked
Gene Aliases: DXS1272E; JARID1C; KDM5C; MRX13; MRXJ; MRXSCJ; MRXSJ; SMCX; XE169
UniProt ID: (Human) P41229
Entrez Gene ID: (Human) 8242
Molecular Function:
histone modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support