Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: FEEGPSRKRP RLQAQRKFAQ SQPNSPSTTP VKIVEPLLPP PATQISDLSK RKPKTEDFLT FLCLRGSPAL PNSMVYFGSS QDEEEVEEED DETEDVKTAT N
Jumonji (JARID2) is a nuclear protein that participates in the negative regulation of cell growth. It is required for neural tube formation and is essential for normal heart development and function. Also, it is a part of a group of proteins characterized by a novel structural motif, the JmjC domain, which is implicated in chromatin regulation through histone demethylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: JARD2 antibody; jumonji homolog; jumonji, AT rich interactive domain 2; jumonji-like protein; Jumonji/ARID domain-containing protein 2; Protein Jumonji
Gene Aliases: JARID2; JMJ; jumonji
UniProt ID: (Human) Q92833, (Mouse) Q62315
Entrez Gene ID: (Human) 3720, (Mouse) 16468, (Rat) 681740
Molecular Function:
histone modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support