Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MTGIAAASFF SNTCRFGGCG LHFPTLADLI EHIEDNHIDT DPRVLEKQEL QQPTYVALSY INRFMTDAAR REQESLKKKI QPKLSLTLSS SVSRGNVSTP PRHSSGSLTP PVTPPITPSS SFRSSTPTGS EYGEEEVDYE ESDSDESWTT ESAISSEAIL SSMCMNGGEE KPFACPVPGC KKRYKNVNGI KYHAKNGHRT QIRVRKPFKC RCGKSYKTAQ GLRHHTINFH PPVSAEIIRK MQQ
JAZF1 is a nuclear protein with three C2H2-type zinc fingers, and functions as a transcriptional repressor. Chromosomal aberrations involving this gene are associated with endometrial stromal tumors.This gene encodes a nuclear protein with three C2H2-type zinc fingers, and functions as a transcriptional repressor. Chromosomal aberrations involving this gene are associated with endometrial stromal tumors. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp761K2222; juxtaposed with another zinc finger gene 1; Juxtaposed with another zinc finger protein 1; TAK1-interacting protein 27; Zinc finger protein 802
Gene Aliases: JAZF1; TIP27; ZNF802
UniProt ID: (Human) Q86VZ6
Entrez Gene ID: (Human) 221895
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support