Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSEKEAMMMV EPHQKVAWKR AVRGVREMCD VCETTLFNIH WVCRKCGFGV CLDCYRLRKS RPRSETEEMG DEEVFSWLKC AKGQSHEPEN LMPTQIIPGT ALYNIGDMVH AARGKWGIKA NCPCISRQNK SVLRPAVTNG MSQLPSINPS ASSGNETTFS GGGGPAPVTT PEPDHVPKAD STDIRSEEPL KTDSSASNSN SELKAIRPPC PDTAPPSSAL HWLADLATQK AKEETKEAGS LRSVLNKESH SPFGLDSFNS TAKVSPLTPK LFNSLLLGPT ASNNKTEGSS LRDLLHSGPG KLPQTPLDTG IPFPPVFSTS SAGVKSKASL PNFLDHIIAS VVENKKTSDA SKRACNLTDT QKEVKEMVMG LNVLDPHTSH SWLCDGRLLC LHDPSNKNNW KIFRECWKQG QPVLVSGVHK KLKSELWKPE AFSQEFGDQD VDLVNCRNCA IISDVKVRDF WDGFEIICKR LRSEDGQPMV LKLKDWPPGE DFRDMMPTRF EDLMENLPLP EYTKRDGRLN LASRLPSYFV RPDLGPKMYN AYGLITAEDR RVGTTNLHLD VSDAVNVMVY VGIPIGEGAH DEEVLKTIDE GDADEVTKQR IHDGKEKPGA LWHIYAAKDA EKIRELLRKV GEEQGQENPP DHDPIHDQSW YLDQTLRKRL YEEYGVQGWA IVQFLGDAVF IPAGAPHQVH NLYSCIKVAE DFVSPEHVKH CFRLTQEFRH LSNTHTNHED KLQVKNIIYH AVKDAVGTLK AHESKLARS
Histone demethylase that specifically demethylates 'Lys-9' of histone H3, thereby playing a central role in histone code. Demethylation of Lys residue generates formaldehyde and succinate. May have tumor suppressor activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [histone H3]-dimethyl-L-lysine(9) demethylase 3B; JmjC domain-containing histone demethylation protein 2B; jumonji domain containing 1B; Jumonji domain-containing protein 1B; lysine (K)-specific demethylase 3B; Lysine-specific demethylase 3B; Nuclear protein 5qNCA
Gene Aliases: 5qNCA; C5orf7; JHDM2B; JMJD1B; KDM3B; KIAA1082; NET22
UniProt ID: (Human) Q7LBC6
Entrez Gene ID: (Human) 51780
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support