Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: METMKSKANC AQNPNCNIMI FHPTKEEFND FDKYIAYMES QGAHRAGLAK IIPPKEWKAR ETYDNISEIL IATPLQQVAS GRAGVFTQYH KKKKAMTVGE YRHLANSKKY QTPPHQNFED LERKYWKNRI YNSPIYGADI SGSLFDENTK QWNLGHLGTI QDLLEKECGV VIEGVNTPYL YFGMWKTTFA WHTEDMDLYS INYLHLGEPK TWYVVPPEHG QRLERLAREL FPGSSRGCGA FLRHKVALIS PTVLKENGIP FNRITQEAGE FMVTFPYGYH AGFNHGFNCA EAINFATPRW IDYGKMASQC SCGEARVTFS MDAFVRILQP ERYDLWKRGQ DRAVVDHMEP RVPASQELST QKEVQLPRRA ALGLRQLPSH WARHSPWPMA ARSGTRCHTL VCSSLPRRSA VSGTATQPRA AAVHSSKKPS STPSSTPGPS AQIIHPSNGR RGRGRPPQKL RAQELTLQTP AKRPLLAGTT CTASGPEPEP LPEDGALMDK PVPLSPGLQH PVKASGCSWA PVP
JMJD2D belongs to JMJD2 family that is consist of four cancer-associated genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [histone H3]-trimethyl-L-lysine(9) demethylase 4D; FLJ10251; JmjC domain-containing histone demethylation protein 3D; jumonji domain containing 2D; Jumonji domain-containing protein 2D; lysine (K)-specific demethylase 4D; Lysine-specific demethylase 4D
Gene Aliases: JHDM3D; JMJD2D; KDM4D
UniProt ID: (Human) Q6B0I6
Entrez Gene ID: (Human) 55693
Molecular Function:
histone modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support