Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene likely encodes a histone lysine demethylase. Studies of a similar protein in mouse indicate a potential role for this protein as a tumor suppressor. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bifunctional peptidase and arginyl-hydroxylase JMJD5; JmjC domain-containing protein 5; Jumonji C domain-containing protein 5; jumonji domain containing 5; jumonji domain-containing protein 5; L-arginine (3R)-hydroxylase KDM8; lysine (K)-specific demethylase 8; Lysine-specific demethylase 8
Gene Aliases: 3110005O21Rik; JMJD5; KDM8; RGD1304823
UniProt ID: (Human) Q8N371, (Mouse) Q9CXT6, (Rat) Q497B8
Entrez Gene ID: (Human) 79831, (Mouse) 77035, (Rat) 308976
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support