Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNHKSKKRI REAKRSARPE LKDSLDWTRH NYYESFSLSP AAVADNVERA DALQLSVEEF VERYERPYKP VVLLNAQEGW SAQEKWTLER LKRKYRNQKF KCGEDNDGYS VKMKMKYYIE YMESTRDDSP LYIFDSSYGE HPKRRKLLED YKVPKFFTDD LFQYAGEKRR PPYRWFVMGP PRSGTGIHID PLGTSAWNAL VQGHKRWCLF PTSTPRELIK VTRDEGGNQQ DEAITWFNVI YPRTQLPTWP PEFKPLEILQ KPGETVFVPG GWWHVVLNLD TTIAITQNFA SSTNFPVVWH KTVRGRPKLS RKWYRILKQE HPELAVLADS VDLQESTGIA SDSSSDSSSS SSSSSSDSDS ECESGSEGDG TVHRRKKRRT CSMVGNGDTT SQDDCVSKER SSSR (1-403 aa encoded by BC066654)
This gene encodes a nuclear protein with a JmjC domain. JmjC domain-containing proteins are predicted to function as protein hydroxylases or histone demethylases. This protein was first identified as a putative phosphatidylserine receptor involved in phagocytosis of apoptotic cells; however, subsequent studies have indicated that it does not directly function in the clearance of apoptotic cells, and questioned whether it is a true phosphatidylserine receptor. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: apoptotic cell clearance receptor PtdSerR; Bifunctional arginine demethylase and lysyl-hydroxylase JMJD6; Histone arginine demethylase JMJD6; JmjC domain-containing protein 6; jumonji domain containing 6; Jumonji domain-containing protein 6; Lysyl-hydroxylase JMJD6; Peptide-lysine 5-dioxygenase JMJD6; Phosphatidylserine receptor; Protein PTDSR
Gene Aliases: 5730436I23Rik; D11Ertd195e; JMJD6; KIAA0585; mKIAA0585; PSR; PtdSerR; PTDSR; PTDSR1
UniProt ID: (Human) Q6NYC1, (Mouse) Q9ERI5, (Rat) Q6AYK2
Entrez Gene ID: (Human) 23210, (Mouse) 107817, (Rat) 360665
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support