Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive Samples: Mouse brain, Mouse testis
Immunogen sequence: CGIKHLHSAG IIHRDLKPSN IVVKSDCTLK ILDFGLARTA GTSFMMTPYV VTRYYRAPEV ILGMGYKENV DIWSVGCIMG EMVRHKILFP GRDYIDQWNK VIEQLGTPCP EFMKKLQPTV RNYVENRPKY AGLTFPKLFP DSLFPADSEH NKLKASQARD LLSKMLVIDP AKRISVDDAL QHPYINVWYD PAEVEAPPPQ IYDKQLDERE HTIEEWKELI YKEVMNSEEK TKNGVVKGQP SPSGAAVNSS ESLPPSSSVN DISSMSTDQT LASDTDSSLE ASAGPLGCCR
MAPK10 (JNK3) is a neuronal-specific form of c-Jun N-terminal kinases (JNKs). MAPK10 is involved in neuronal apoptosis and phosphorylates MAP2K4. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. This protein is a neuronal-specific form of c-Jun N-terminal kinases (JNKs). Through its phosphorylation and nuclear localization, this kinase plays regulatory roles in the signaling pathways during neuronal apoptosis. Beta-arrestin 2, a receptor-regulated MAP kinase scaffold protein, is found to interact with, and stimulate the phosphorylation of this kinase by MAP kinase kinase 4 (NhpK4). Cyclin-dependent kianse 5 can phosphorylate, and inhibit the activity of this kinase, which may be important in preventing neuronal apoptosis. Four alternatively spliced transcript variants encoding distinct isoforms have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-Jun N-terminal kinase 3; JNK3 beta1 protein kinase; JNK3 beta2 protein kinase; MAP kinase 10; MAP kinase p49 3F12; MAPK 10; mitogen activated protein kinase 10; Mitogen-activated protein kinase 10; OTTHUMP00000161181; OTTHUMP00000161183; OTTHUMP00000218718; SAPK/Erk/kinase 2; Stress-activated protein kinase JNK3
Gene Aliases: C230008H04Rik; JNK3; JNK3B1; JNK3B2; Mapk10; p493F12; p54bSAPK; Prkm10; SAPK(beta); Serk2
UniProt ID: (Mouse) Q61831
Entrez Gene ID: (Mouse) 26414
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support