Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
JTV1 is probable core protein of the multisynthetase complex that serves as a template for the assembly of the supramolecular structure. This protein Mediates ubiquitination of FUBP1 and its degradation by the proteasome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aminoacyl tRNA synthase complex-interacting multifunctional protein 2; ARS interacting multifunctional protein 2; ARS-interacting multi-functional protein 2; JTV1; Multisynthase complex auxiliary component p38; multisynthetase complex auxiliary component p38; Protein JTV-1
Gene Aliases: AA407136; AA407225; AIMP2; Aimp2(p38); JTV-1; JTV1; P38; PRO0992
UniProt ID: (Human) Q13155, (Rat) Q32PX2, (Mouse) Q8R010
Entrez Gene ID: (Human) 7965, (Rat) 288480, (Mouse) 231872
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support