Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MERLGEKASR LLEKFGRRKG ESSRSGSDGT PGPGKGRLSG LGGPRKSGPR GATGGPGDEP LEPAREQGSL DAERNQRGSF EAPRYEGSFP AGPPPTRALP LPQSLPPDFR LEPTAPALSP RSSFASSSAS DASKPSSPRG SLLLDGAGAG GAGGSRPCSN RTSGISMGYD QRHGSPLPAG PCLFGPPLAG APAGYSPGGV PSAYPELHAA LDRLYAQRPA GFGCQESRHS YPPALGSPGA LAGAGVGAAG PLERRGAQPG RHSVTGYGDC AVGARYQDEL TALLRLTVGT GGREAGARGE PSGIEPSGLE EPPGPFVPEA ARARMREPEA REDYFGTCIK CNKGIYGQSN ACQALDSLYH TQCFVCCSCG RTLRCKAFYS VNGSVYCEED YLFSGFQEAA EKCCVCGHLI LEKILQAMGK SYHPGCFRCI VCNKCLDGIP FTVDFSNQVY CVTDYHKNYA PKCAACGQPI LPSEGCEDIV RVISMDRDYH FECYHCEDCR MQLSDEEGCC CFPLDGHLLC HGCHMQRLNA RQPPANYI
AJUBA (Ajuba LIM Protein) is a Protein Coding gene. Diseases associated with AJUBA include Wilson-Turner X-Linked Mental Retardation Syndrome and Syndromic X-Linked Intellectual Disability. Among its related pathways are Hippo signaling pathway and Adhesion. Gene Ontology (GO) annotations related to this gene include chromatin binding and actin filament binding. An important paralog of this gene is LIMD1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: jub, ajuba homolog; LIM domain-containing protein ajuba
Gene Aliases: AJUBA; JUB
UniProt ID: (Human) Q96IF1
Entrez Gene ID: (Human) 84962
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support