Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Sequence of this protein is as follows: MCTKMEQPFY HDDSYTATGY GRAPGGLSLH DYKLLKPSLA VNLADPYRSL KAPGARGPGP EGGGGGSYFS GQGSDTGASL KLASSELERL IVPNSNGVIT TTPTPPGQYF YPRGGGSGGG AGGAGGGVTE EQEGFADGFV KALDDLHKMN HVTPPNVSLG ATGGPPAGPG GVYAGPEPPP VYTNLSSYSP ASASSGGAGA AVGTGSSYPT TTISYLPHAP PFAGGHPAQL GLGRGASTFK EEPQTVPEAR SRDATPPVSP INMEDQERIK VERKRLRNRL AATKCRKRKL ERIARLEDKV KTLKAENAGL SSTAGLLREQ VAQLKQKVMT HVSNGCQLLL GVKGHAF
JunB, a member of the Jun cellular oncogene family, is a basic region, leucine zipper transcription factor. It is involved in regulating gene activity following the primary growth factor response. JunB binds to the DNA sequence 5'-TGA[CG]TCA-3'. Cellular oncogenes play critical roles in cellular communication pathways that regulate growth, development, and differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: activator protein 1; Jun B; jun B proto-oncogene; Oncogene Jun-B; Transcription factor AP-1 subunit JunB; Transcription factor JunB
Gene Aliases: AP-1; JUNB
UniProt ID: (Human) P17275
Entrez Gene ID: (Human) 3726
Molecular Function:
basic leucine zipper transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support