Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
The Notch signaling pathway is an intercellular signaling mechanism that is essential for proper embryonic development. Members of the Notch gene family encode transmembrane receptors that are critical for various cell fate decisions. JAG2 is one of several ligands that activate Notch and related receptors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Jagged2; Protein jagged-2; syndactylism
Gene Aliases: D12Ggc2e; HJ2; JAG2; mJagged2-1; SER2; Serh; sm
UniProt ID: (Human) Q9Y219, (Mouse) Q9QYE5
Entrez Gene ID: (Human) 3714, (Mouse) 16450, (Rat) 29147
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support