Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
The jagged 1 protein encoded by JAG1 is the human homolog of the Drosophilia jagged protein. Human jagged 1 is the ligand for the receptor notch 1, the latter a human homolog of the Drosophilia jagged receptor notch. Mutations that alter the jagged 1 protein cause Alagille syndrome. Jagged 1 signalling through notch 1 has also been shown to play a role in hematopoiesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD339; jagged canonical Notch ligand 1; Jagged1; Protein jagged-1; Serrate-1
Gene Aliases: ABE2; AGS; AHD; AWS; CD339; Gsfabe2; HJ1; Htu; JAG1; JAGL1; Ozz; Ser-1
UniProt ID: (Human) P78504, (Mouse) Q9QXX0, (Rat) Q63722
Entrez Gene ID: (Human) 182, (Mouse) 16449, (Rat) 29146
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support