Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Positive test controls include: HeLa, HT-1080. The target is usually found in the following locations: Nucleus.
Immunogen sequence: SPELERLIIQ SNGLVTTTPT SSQFLYPKVA ASEEQEFAEG FVKALEDLHK QNQLGAGAAA AAAAAAAGGP SGTATGSAPP GELAPAAAAP EAPVYANLSS Y
JunD is a transcription factor which is coded by JunD gene. JunD transcriptional activity is modulated by phosphorylation in response to cellular stress via the c-Jun N terminal kinase (JNK)/Stress-Activated Protein Kinase (SAPK) family of protein kinase. Jun proteins are capable of forming dimers with Fos, ATF and CREB family transcription factors to form the AP-1 complex that differentially regulates a variety of target genes involved in cellular growth, proliferation, differentiation and apoptosis. Also, JunD plays an important role in metabolism via modulation of IGF-1 signaling pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: activator protein 1; JUN D; jun D proto-oncogene; Jun proto-oncogene related gene d; Jun proto-oncogene related gene d1; JunD-FL isoform; Oncogene Jun-D; Transcription factor AP-1 subunit JunD; transcription factor jun-D; Transcription factor JunD
Gene Aliases: AP-1; Jun-d; JUND; Jund1
UniProt ID: (Human) P17535, (Rat) P52909, (Mouse) P15066
Entrez Gene ID: (Human) 3727, (Rat) 24518, (Mouse) 16478
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support