Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody can recognize isoform1 and isoform2 of KALRN.
HAP1 (huntingtin-associated protein 1) binds to huntingtin. Huntingtin is a protein that contains a polyglutamine region and when the number of glutamine repeats exceeds 35, the gene encodes a version of huntingtin that leads to Huntington's disease (HD). The ability of HAP1 to bind to huntingtin is enhanced by an expanded polyglutamine repeat region. HAP1 shows neuronal localization and moves with huntingtin in nerve fibers. HAP1 is primarily expressed in brain tissue, with greater expression in the olfactory bulb and brain stem. Mouse HAP1 is localized to membrane-bound organelles including large endosomes, tubulovesicular structures and budding vesicles in neurons. Duo, also designated huntingtin-associated protein interacting protein or HAPIP, binds Huntingtin-associated protein 1 (HAP1) and may have a role in vesicle trafficking and cytoskeletal function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ12332; FLJ16443; FLJ18196; FLJ18623; hCG_2022551; huntingtin-associated protein interacting protein (duo); Huntingtin-associated protein-interacting protein; Kalirin; kalrn; Protein Duo; RhoGEF kinase; serine/threonine kinase with Dbl- and pleckstrin homology domain; serine/threonine kinase with Dbl- and pleckstrin homology domains; Serine/threonine-protein kinase with Dbl- and pleckstrin homology domain
Gene Aliases: 2210407G14Rik; ARHGEF24; AV235988; CHD5; CHDS5; DUET; DUO; E530005C20Rik; Gm539; HAPIP; KALRN; TRAD
UniProt ID: (Human) O60229, (Mouse) A2CG49
Entrez Gene ID: (Human) 8997, (Mouse) 545156
Molecular Function:
guanyl-nucleotide exchange factor
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support