Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SQHIPTAEGA SGSNTEEEIR MELSPDLISA CLALEKYLDN PNALTERELK VAYTTVLQEW LRLACRSDAH PELVRRHLVT FRAMSARLLD YVVNIADSNG NTALHYSVSH ANFPVVQ
Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.
This gene encodes a member of the KN motif and ankyrin repeat domains (KANK) family of proteins, which play a role in cytoskeletal formation by regulating actin polymerization. The encoded protein functions in the sequestration of steroid receptor coactivators and possibly other proteins. Mutations in this gene are associated with impaired kidney podocyte function and nephrotic syndrome, and keratoderma and woolly hair.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ankyrin repeat domain-containing protein 25; kidney ankyrin repeat-containing protein 2; KN motif and ankyrin repeat domain-containing protein 2; Matrix-remodeling-associated protein 3; SIP; SRC-1-interacting protein; SRC-interacting protein; SRC1-interacting protein
Gene Aliases: ANKRD25; KANK2; KIAA1518; MXRA3; PPKWH; SIP
UniProt ID: (Human) Q63ZY3
Entrez Gene ID: (Human) 25959
Molecular Function:
non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support