Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ALIMREKLI RLQSLYIDQF KRLQHLLKEK KRRYLHNRKV EHEALGSSLL TGPEGLLAKE RENLKRLKCL RRYRQRYGVE ALLHRQLKER RMLATDGAAQ QAHTTRSSQR CLAFVDDVRC SNQSLPMTRH CLTHICQDTN QVLFKCCQGS EEVPCNKPVP VSLSEDPCCP LHFQLPPQMY KPEQVLSVPD DLEAGPMDLY LSAAELQPTE SLPLEFSDDL DVVGDGMQCP PSPLLFDPSL TLEDHLVKEI AEDTVDILGQ MQMAGDGCRS QGSRNSEKAS APLSQSGLAT ANGKPEPTSI S (193-492 aa encoded by BC009746)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KAT8 regulatory NSL complex subunit 2; Non-specific lethal 2 homolog; NSL complex protein NSL2
Gene Aliases: 2310037I24Rik; B930009D17; C12orf41; KANSL2; NSL2
UniProt ID: (Human) Q9H9L4, (Mouse) Q8BQR4
Entrez Gene ID: (Human) 54934, (Mouse) 69612
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support