Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MNLAEICDNA KKGREYALLG NYDSSMVYYQ GVMQQIQRHC QSVRDPAIKG KWQQVRQELL EEYEQVKSIV STLESFKIDK PPDFPVSCQD EPFRDPAVWP PPVPAEHRAP PQIRRPNREV RPLRKEMAGV GARGPVGRAH PISKSEKPST SRDKDYRARG RDDKGRKNMQ DGASDGEMPK FDGAGYDKDL VEALERDIVS RNPSIHWDDI ADLEEAKKLL REAVVLPMWM PDFFKGIRRP WKGVLMVGPP GTGKTMLAKA VATECGTTFF NVSSSTLTSK YRGESEKLVR LLFEMARFYA PTTIFIDEID SICSRRGTSD EHEASRRVKS ELLIQMDGVG GALENDDPSK MVMVLAATNF PWDIDEALRR RLEKRIYIPL PTAKGRAELL KINLREVELD PDIQLEDIAE KIEGYSGADI TNVCRDASLM AMRRRINGLS PEEIRALSKE ELQMPVTKGD FELALKKIAK SVSAADLEKY EKWMVEFGSA
KATNAL1 (Katanin Catalytic Subunit A1 Like 1) is a Protein Coding gene. Diseases associated with KATNAL1 include 13Q12.3 Microdeletion Syndrome. Gene Ontology (GO) annotations related to this gene include microtubule binding and microtubule-severing ATPase activity. An important paralog of this gene is KATNA1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: katanin catalytic subunit A like 1; Katanin p60 ATPase-containing subunit A-like 1; katanin p60 subunit A like 1; Katanin p60 subunit A-like 1; KATNAL1; p60 katanin-like 1
Gene Aliases: KATNAL1
UniProt ID: (Human) Q9BW62
Entrez Gene ID: (Human) 84056
Molecular Function:
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support