Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
KATNAL1 (Katanin Catalytic Subunit A1 Like 1) is a Protein Coding gene. Diseases associated with KATNAL1 include 13Q12.3 Microdeletion Syndrome. Gene Ontology (GO) annotations related to this gene include microtubule binding and microtubule-severing ATPase activity. An important paralog of this gene is KATNA1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: katanin catalytic subunit A like 1; Katanin p60 ATPase-containing subunit A-like 1; katanin p60 ATPase-containing subunit A-like 1 {ECO:0000255|HAMAP-Rule:MF_03024}; katanin p60 subunit A like 1; Katanin p60 subunit A-like 1; katanin p60 subunit A-like 1 {ECO:0000255|HAMAP-Rule:MF_03024}; KATNAL1; p60 katanin-like 1; p60 katanin-like 1 {ECO:0000255|HAMAP-Rule:MF_03024}
Gene Aliases: KATNAL1
UniProt ID: (Human) Q9BW62, (Rat) Q5XIK7
Entrez Gene ID: (Human) 84056, (Rat) 288449
Molecular Function:
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support