Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
KATNAL1 (Katanin Catalytic Subunit A1 Like 1) is a Protein Coding gene. Diseases associated with KATNAL1 include 13Q12.3 Microdeletion Syndrome. Gene Ontology (GO) annotations related to this gene include microtubule binding and microtubule-severing ATPase activity. An important paralog of this gene is KATNA1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: katanin catalytic subunit A like 1; Katanin p60 ATPase-containing subunit A-like 1; katanin p60 subunit A like 1; Katanin p60 subunit A-like 1; KATNAL1; p60 katanin-like 1
Gene Aliases: KATNAL1
UniProt ID: (Human) Q9BW62
Entrez Gene ID: (Human) 84056
Molecular Function: non-motor microtubule binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support