Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLPPPRPAAA LALPVLLLLL VVLTPPPTGA RPSPGPDYLR RGWMRLLAEG EGCAPCRPEE CAAPRGCLAG RVRDACGCCW ECANLEGQLC DLDPSAHFYG HCGEQLECRL DTGGDLSRGE VPEPLCACRS QSPLCGSDGH TYSQICRLQE AARARPDANL TVAHPGPCES GPQIVSHPYD TWNVTGQDVI FGCEVFAYPM ASIEWRKDGL DIQLPGDDPH ISVQFRGGPQ RFEVTGWLQI QAVRPSDEGT YRCLGRNALG QVEAPASLTV LTPDQLNSTG IPQLRSLNLV PEEEAESEEN DDYY
This gene encodes a secreted member of the insulin growth factor-binding protein (IGFBP) superfamily. It contains an N-terminal insulin growth factor-binding domain, a central Kazal-type serine protease inhibitor and follistatin-like domain, and a C-terminal immunoglobulin-like domain. Studies of the mouse ortholog suggest that this gene product may have a function in bone development and bone regeneration.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bone- and odontoblast-expressed gene 1; FLJ24094; IGFBP-related protein 10; Kazal-type serine protease inhibitor domain-containing protein 1; novel kazal-type serine protease inhibitor domain and immunoglobulin domain containing protein
Gene Aliases: BONO1; FKSG28; FKSG40; IGFBP-rP10; KAZALD1; UNQ2945/PRO21184
UniProt ID: (Human) Q96I82
Entrez Gene ID: (Human) 81621
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support