Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Cow: 79%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 79%; Rat: 93%
KBTBD12 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kelch domain containing 6; Kelch domain-containing protein 6; kelch repeat and BTB (POZ) domain containing 12; Kelch repeat and BTB domain-containing protein 12
Gene Aliases: KBTBD12; KLHDC6
UniProt ID: (Human) Q3ZCT8
Entrez Gene ID: (Human) 166348
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support