Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
KBTBD4 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BTB and kelch domain containing 4; BTB and kelch domain-containing protein 4; kelch repeat and BTB (POZ) domain containing 4; Kelch repeat and BTB domain-containing protein 4
Gene Aliases: 2510026C23Rik; BKLHD4; HSPC252; KBTBD4
UniProt ID: (Human) Q9NVX7, (Mouse) Q8R179
Entrez Gene ID: (Human) 55709, (Mouse) 67136
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support