Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TGPEELKDTA HSAALLAQLK SFYDARLLCD VTIEVVTPGS GPGTGRLFPC NRNVLAAACP YFKSM
Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.
KBTBD6 gene ontology (GO) annotation include Cul3-RING ubiquitin ligase complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kelch repeat and BTB (POZ) domain containing 6; Kelch repeat and BTB domain-containing protein 6
Gene Aliases: KBTBD6
UniProt ID: (Human) Q86V97
Entrez Gene ID: (Human) 89890
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support