Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPGHYSLPQP PSQPLSSVVV NMPAQALYAS PQPLAVSTLP GVGQVARPGP TAVGNGHMAG PLLPPPPPAQ PSATLPSGAP ATNGPPTTDS AHGLQMLRTI GVGKYEFTDP GHPREMLKEL NQQRRAKAFT DLKIVVEGRE FEVHQNVLAS CSLYFKDLIQ RSVQDSGQGG REKLELVLSN LQADVLELLL EFVYTGSLVI DSANAKTLLE AASKFQFHTF CKVCVSFLEK QLTASNCLGV LAMAEAMQCS ELYHMAKAFA LQIFPEVAAQ EEILSISKDD FIAYVSNDSL NTKAEELVYE TVIKWIKKDP ATRTQYAAEL LAVVRLPFIH PSYLLNVVDN EELIKSSEAC RDLVNEAKRY HMLPHARQEM QTPRTRPRLS AGVAEVIVLV GGRQMVGMTQ RSLVAVTCWN PQNNKWYPLA SLPFYDREFF SVVSAGDNIY LSGGMESGVT LADVWCYMSL LDNWNLVSRM TVPRCRHNSL VYDGKIYTLG GLGVAGNVDH VER
KLHL29 (Kelch-like protein 29), also known as KBTBD9, is a 655 amino acid protein that is related to the Drosophila Kelch protein. Mutations affecting Kelch function result in failure of Kelch to associate with the ring canals and subsequent female sterility. Human KLHL29 contains six kelch repeats and one BTB (POZ) domain. The BTB (Broad-Complex, Tramtrack and Bric a brac) domain, also known as the POZ (Poxvirus and Zinc finger) domain, is an N-terminal homodimerization domain that contains multiple copies of kelch repeats and/or C2H2-type zinc fingers. Proteins that contain BTB domains are thought to be involved in transcriptional regulation via control of chromatin structure and function. KLHL29 exists as two alternatively spliced isoforms which are encoded by a gene that maps to human chromosome 2p24.1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kelch repeat and BTB (POZ) domain containing 9; Kelch repeat and BTB domain-containing protein 9; kelch-like 29; kelch-like family member 29; Kelch-like protein 29
Gene Aliases: KBTBD9; KIAA1921; KLHL29
UniProt ID: (Human) Q96CT2
Entrez Gene ID: (Human) 114818
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support