Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Reconstitution: 1 X 25 µL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
SLC12A4 mediates electroneutral potassium-chloride cotransport when activated by cell swelling. SLC12A4 may contribute to cell volume homeostasis in single cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Electroneutral potassium-chloride cotransporter 1; Erythroid K-Cl cotransporter 1; FLJ40489; Furosemide-sensitive K-Cl cotransporter; hKCC1; K-Cl Co-transporter-1; K-Cl cotransporter KCC1; mKCC1; potassium/chloride cotransporter 1; rKCC1; solute carrier family 12 (potassium/chloride transporter), member 4; solute carrier family 12 (potassium/chloride transporters), member 4; Solute carrier family 12 member 4; solute carrier family 12, member 4
Gene Aliases: AW546649; CTC-479C5.17; hKCC1; KCC1; RBCKCC1; SLC12A4
UniProt ID: (Human) Q9UP95, (Mouse) Q9JIS8, (Rat) Q63632
Entrez Gene ID: (Human) 6560, (Mouse) 20498, (Rat) 29501
Molecular Function:
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support