Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
Mediates electroneutral potassium-chloride cotransport in mature neurons. Transport occurs under isotonic conditions, but is activated 20-fold by cell swelling. Important for Cl(-) homeostasis in neurons.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Electroneutral potassium-chloride cotransporter 2; erythroid K-Cl cotransporter 2; Furosemide-sensitive K-Cl cotransporter; hKCC2; K-Cl cotransporter 2; KCC 2; KCC2a variant 1; KCC2a-S25 variant 1; KCC2b variant 2; mKCC2; Neuronal K-Cl cotransporter; neuronal-specific K-Cl cotransporter; rKCC2; S12A5; solute carrier family 12 (potassium/chloride transporter), member 5; Solute carrier family 12 member 5; solute carrier family 12, member 5
Gene Aliases: EIEE34; EIG14; hKCC2; KCC2; KIAA1176; mKIAA1176; SLC12A5
UniProt ID: (Human) Q9H2X9, (Mouse) Q91V14, (Rat) Q63633
Entrez Gene ID: (Human) 57468, (Mouse) 57138, (Rat) 171373
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support