Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
Immunogen sequence: STVKSKRREH SNDPYHQYIV EDWQEKYKSQ ILNLEESKAT IHENIGAAGF KMS
Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 83%.
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, isk-related subfamily. This member is a small integral membrane subunit that assembles with the KCNH2 gene product, a pore-forming protein, to alter its function. This gene is expressed in heart and muscle and the gene mutations are associated with cardiac arrhythmia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cardiac voltage-gated potassium channel accessory subunit 2; CDW210B; CRFB4; D21S58; D21S66; IL-10R2; Minimum potassium ion channel-related peptide 1; MinK-related peptide 1; minK-related peptide-1; Potassium channel subunit beta MiRP1; potassium channel subunit, MiRP1; potassium channel, voltage gated subfamily E regulatory beta subunit 2; Potassium voltage-gated channel subfamily E member 2; potassium voltage-gated channel, Isk-related family, member 2; voltage-gated K+ channel subunit MIRP1
Gene Aliases: ATFB4; KCNE2; LQT5; LQT6; MIRP1
UniProt ID: (Human) Q9Y6J6
Entrez Gene ID: (Human) 9992
Molecular Function: voltage-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support