Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPVRRGHVAP QNTYLDTIIR KFEGQSRKFL IANAQMENCA IIYCNDGFCE LFGYSRVEVM QQPCTCDFLT GPNTPSSAVS RLAQALLGAE ECKVDILYYR KDASSFRCLV DVVPVKNEDG AVIMFILNFE DLAQLLAKCS SRSLSQRLLS QSFLGSEGSH GRPGGPGPGT GRGKYRTISQ IPQFTLNFVE FNLEKHRSSS TTEIEIIAPH KVVERTQNVT EKVTQVLSLG ADVLPEYKLQ APRIHRWTIL HYSPFKAVWD WLILLLVIYT AVFTPYSAAF LLSDQDESRR GACSYTCSPL TVVDLIVDIM FVVDIVINFR TTYVNTNDEV VSHPRRIAVH YFKGWFLIDM VAAIPFDLLI FRTGSDETTT LIGLLKTARL LRLVRVARKL DRYSEYGAAV LFLLMCTFAL IAHWLACIWY AIGNVERPYL EHKIGWLDSL GVQLGKRYNG SDPASGPSVQ DKYVTALYFT FSSLTSVGFG NVSPNTNSEK VFSICVMLIG CE
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. KCNH6 encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eag-related gene member 2; ERG-2; Ether-a-go-go-related gene potassium channel 2; ether-a-go-go-related protein 2; potassium channel, voltage gated eag related subfamily H, member 6; Potassium voltage-gated channel subfamily H member 6; potassium voltage-gated channel, subfamily H (eag-related), member 6; Voltage-gated potassium channel subunit Kv11.2
Gene Aliases: ERG-2; ERG2; hERG-2; HERG2; KCNH6; Kv11.2
UniProt ID: (Human) Q9H252
Entrez Gene ID: (Human) 81033
Molecular Function:
ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support