Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MGAVMGTFSS LQTKQRRPSK DKIEDELEMT MVCHRPEGLE QLEAQTNFTK RELQVLYRGF KNECPSGVVN EDTFKQIYAQ FFPHGDASTY AHYLFNAFDT TQTGSVKFED FVTALSILLR GTVHEKLRWT FNLYDINKDG YINKEEMMDI VKAIYDMMGK YTYPVLKEDT PRQHVDVFFQ KMDKNKDGIV TLDEFLESCQ EDDNIMRSLQ LFQNVM
This gene encodes a member of the family of voltage-gated potassium (Kv) channel-interacting proteins (KCNIPs), which belong to the recoverin branch of the EF-hand superfamily. Members of the KCNIP family are small calcium binding proteins. They all have EF-hand-like domains, and differ from each other in the N-terminus. They are integral subunit components of native Kv4 channel complexes. They may regulate A-type currents, and hence neuronal excitability, in response to changes in intracellular calcium. Alternative splicing results in multiple transcript variant encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A-type potassium channel modulatory protein 1; KChIP1; KCHIP2; Kv channel interacting protein 1; Kv channel-interacting protein 1; potassium channel interacting protein 1; Potassium channel-interacting protein 1; Vesicle APC-binding protein
Gene Aliases: KCHIP1; KCNIP1; VABP
UniProt ID: (Human) Q9NZI2
Entrez Gene ID: (Human) 30820
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support