Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ILAKISRPK KRAKTITFSK NAVISKRGGK LCLLIRVANL RKSLLIGSHI YGKLLKTTVT PEGETIILDQ ININFVVDAG NENLFFISPL TIYHVIDHNS PFFHMAAETL LQQDFELVVF LDGTVESTSA TCQVRTSYVP EEVLWGYRFA PIVSKTKEGK YRVDFHNFSK TVEVETPHCA MCLYNEKDVR ARMKRGYDNP NFILSEVNET DDTKM (178-391 aa encoded by BC074752)
Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. It is activated by internal ATP and probably plays an important role in potassium homeostasis. The encoded protein has a greater tendency to allow potassium to flow into a cell rather than out of a cell. Mutations in this gene have been associated with antenatal Bartter syndrome, which is characterized by salt wasting, hypokalemic alkalosis, hypercalciuria, and low blood pressure. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-regulated potassium channel ROM-K; ATP-sensitive inward rectifier potassium channel 1; Inward rectifier K(+) channel Kir1.1; inwardly rectifying K+ channel; inwardly rectifying potassium channel ROMK-2; K+ channel protein; KAB-1; kir1.1; OTTHUMP00000045938; Potassium channel, inwardly rectifying subfamily J member 1; potassium channel, inwardly rectifying subfamily J, member 1; Potassium inwardly-rectifying channel subfamily J; potassium inwardly-rectifying channel, subfamily J, member 1
Gene Aliases: Kcnj; KCNJ1; KIR1.1; ROMK; ROMK1; Romk2
UniProt ID: (Human) P48048, (Mouse) O88335
Entrez Gene ID: (Human) 3758, (Mouse) 56379, (Rat) 24521
Molecular Function:
ion channel
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support