Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
KCNK3 encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. The gene product is an outwardly rectifying channel that is sensitive to changes in extracellular pH and is inhibited by extracellular acidification. Also referred to as an acid-sensitive potassium channel, it is activated by the anesthetics halothane and isoflurane. Although three transcripts are detected in northern blots, there is currently no sequence available to confirm transcript variants for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acid-sensitive potassium channel protein TASK; Acid-sensitive potassium channel protein TASK-1; cardiac potassium channel; Cardiac two pore background K(+) channel; cTBAK 1; cTBAK-1; KCNK9; Potassium channel subfamily K member 3; potassium channel, subfamily K, member 3; potassium channel, two pore domain subfamily K, member 3; potassium inwardly-rectifying channel, subfamily K, member 3; TASK 1; TWIK-related acid-sensitive K(+) channel 1; TWIK-related acid-sensitive K+ channel; two P domain potassium channel; Two pore K(+) channel KT3.1; Two pore potassium channel KT3.1
Gene Aliases: Ctbak; cTBAK-1; K2p3.1; KCNK3; OAT1; PPH4; rTASK; TASK; TASK-1; TASK1; TBAK1
UniProt ID: (Human) O14649, (Rat) O54912, (Mouse) O35111
Entrez Gene ID: (Human) 3777, (Rat) 29553, (Mouse) 16527
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support