Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGGDLVLGLG ALRRRKRLLE QEKSLAGWAL VLAGTGIGLM VLHAEMLWFG GCSWALYLFL VKCTISISTF LLLCLIVAFH AKEVQLFMTD NGLRDWRVAL TGRQAAQIVL ELVVCGLHPA PVRGPPCVQD LGAPLTSPQP WPGFLGQGEA LLSLAMLLRL YLVPRAVLLR SGVLLNASYR SIGALNQVRF RHWFVAKLYM NTHPGRLLLG LTLGLWLTTA WVLSVAERQA VNATGHLSDT LWLIPITFLT IGYGDVVPGT MWGKIVCLCT GVMGVCCTAL LVAVVARKLE FNKAEKHVHN FMMDIQYTKE MKESAARVLQ EAWMFYKHTR RKESHAARRH QRKLLAAINA FRQVRLKHRK LREQVNSMVD ISKMHMILYD LQQNLSSSHR ALEKQIDTLA GKLDALTELL STALGPRQLP EPSQQSK
The protein encoded by this gene is part of a potentially heterotetrameric voltage-independent potassium channel that is activated by intracellular calcium. Activation is followed by membrane hyperpolarization, which promotes calcium influx. The encoded protein may be part of the predominant calcium-activated potassium channel in T-lymphocytes. This gene is similar to other KCNN family potassium channel genes, but it differs enough to possibly be considered as part of a new subfamily.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HIK1; IK1; IKCa; IKCa1; Intermediate conductance calcium-activated potassium channel protein 4; KCa3.1; KCa4; KCNN 4; potassium channel, calcium activated intermediate/small conductance subfamily N alpha, member 4; potassium intermediate/small conductance calcium-activated channel, subfamily N, member 4; putative erythrocyte intermediate conductance calcium-activated potassium Gardos channel; Putative Gardos channel; SK4; SKCa 4; SKCa4; small conductance calcium-activated potassium channel 4
Gene Aliases: DHS2; hIKCa1; hKCa4; hSK4; IK; IK1; IKCA1; KCa3.1; KCA4; KCNN4; SK4
UniProt ID: (Human) O15554
Entrez Gene ID: (Human) 3783
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support