Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Voltage-gated K+ channels in the plasma membrane control the repolarization and the frequency of action potentials in neurons, muscles and other excitable cells. A specific K+ channel, comprised of an alpha subunit KCNQ1 and a beta subunit KCNE1, a small protein which spans the membrane only once, is predominantly expressed in the heart and in the cochlea, and is responsible for regulating the slow, depolarization-activated potassium current. Mutations in the genes encoding for KCNQ1 and KCNE1 lead to cardiac disease because they directly impair electrical signaling, and mutations in KCNQ4 are implicated in the onset of deafness. KCNQ proteins, including KCNQ1 and KCNQ4, characteristically contain six transmembrane domains and function as tetramers. KCNQ4 forms heteromeric channels with KCNQ3 and is expressed in several tissues, including the cochlea, where it is present in outer hair cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ26167; IKs producing slow voltage-gated potassium channel subunit alpha KvLQT1; kidney and cardiac voltage dependend K+ channel; KQT-like 1; potassium channel, voltage gated KQT-like subfamily Q, member 1; Potassium voltage-gated channel subfamily KQT member 1; potassium voltage-gated channel, KQT-like subfamily, member 1; potassium voltage-gated channel, subfamily Q, member 1; slow delayed rectifier channel subunit; Voltage-gated potassium channel subunit Kv7.1
Gene Aliases: ATFB1; ATFB3; AW559127; JLNS1; KCNA8; KCNA9; KCNQ1; Kv1.9; Kv7.1; KVLQT1; LQT; LQT1; RWS; SQT2; WRS
UniProt ID: (Human) P51787, (Mouse) P97414, (Rat) Q9Z0N7
Entrez Gene ID: (Human) 3784, (Mouse) 16535, (Rat) 84020
Molecular Function:
voltage-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support