Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Reconstitution: 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
Voltage-gated potassium channels form the largest and most diversified class of ion channels and are present in both excitable and nonexcitable cells. Their main functions are associated with the regulation of the resting membrane potential and the control of the shape and frequency of action potentials. The alpha subunits are of 2 types: those that are functional by themselves and those that are electrically silent but capable of modulating the activity of specific functional alpha subunits. The protein encoded by this gene is not functional by itself but can form heteromultimers with member 1 and with member 2 (and possibly other members) of the Shab-related subfamily of potassium voltage-gated channel proteins. This gene belongs to the S subfamily of the potassium channel family. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Delayed-rectifier K(+) channel alpha subunit 3; potassium voltage-gated channel delayed-rectifier protein S3; Potassium voltage-gated channel subfamily S member 3; potassium voltage-gated channel, delayed-rectifier, subfamily S, member 3; potassium voltage-gated channel, modifier subfamily S, member 3; Shab-related delayed-rectifier K+ channel (Kv9.3); Shab-related delayed-rectifier K+ channel alpha subunit 3; voltage-gated potassium channel protein Kv9.3; Voltage-gated potassium channel subunit Kv9.3
Gene Aliases: KCNS3; KV9.3
UniProt ID: (Human) Q9BQ31, (Rat) O88759, (Mouse) Q8BQZ8
Entrez Gene ID: (Human) 3790, (Rat) 83588, (Mouse) 238076
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support