Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The antigen is homologous in rat and mouse.
Specificity of this antibody: KCNT2.
Kcnt2 calcium (Ca2+) -activated potassium channels (KCa) are a group of 6/7-TM ion channels that selectively transport K+ ions across biological membranes. KCa channels are broadly classified into three subtypes: SK, IK and BK channels (small, intermediate and big conductance). Kcnt2 is an outward rectifying potassium channel, and produces rapidly activating outward rectifier K(+) currents. Kcnt2 is activated by high intracellular sodium and chloride levels. Further, Kcnt2 channel activity is inhibited by ATP and by inhalation anesthetics, such as isoflurane. Functionally, Kcnt2 is inhibited upon stimulation of G-protein coupled receptors, such as CHRM1 and GRIA1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Potassium channel subfamily T member 2; potassium channel, sodium activated subfamily T, member 2; potassium channel, subfamily T, member 2; Sequence like an intermediate conductance potassium channel subunit; Sodium and chloride-activated ATP-sensitive potassium channel Slo2.1; sodium- and chloride-activated ATP-sensitive potassium channel; sodium- and chloride-activated ATP-sensitive potassium channel subunit; sodium-and chloride-activated ATP-sensitive potassium channel (SLICK)
Gene Aliases: E330038N15Rik; KCa4.2; KCNT2; SLICK; SLO2.1
UniProt ID: (Human) Q3SY59, (Rat) Q6UVM4
Entrez Gene ID: (Human) 343450, (Rat) 304827, (Mouse) 240776
Molecular Function:
ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support