Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: PC-3.
Predicted reactivity: Mouse (82%), Rhesus Monkey (98%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the potassium channel tetramerization-domain containing (KCTD) protein family. Members of this protein family regulate the biophysical characteristics of ion channels. In mouse, this protein interacts with hyperpolarization-activated cyclic nucleotide-gated channel complex 3 and enhances its cell surface expression and current density. Alternative splicing results in multiple transcript variants.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BTB/POZ domain-containing protein KCTD3; NY-REN-45 antigen; potassium channel tetramerisation domain containing 3; Renal carcinoma antigen NY-REN-45
Gene Aliases: KCTD3; NY-REN-45
UniProt ID: (Human) Q9Y597
Entrez Gene ID: (Human) 51133
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support