Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The KCTD gene family, including KCTD7, encode predicted proteins that contain N-terminal domain that is homologous to the T1 domain in voltage-gated potassium channels (see KCNA1; MIM 176260). KCTD7 displays a primary sequence and hydropathy profile indicating intracytoplasmic localization. EST database analysis showed that KCTD7 is expressed in human and mouse brain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BTB/POZ domain-containing protein KCTD7; FLJ32069; potassium channel tetramerisation domain containing 7
Gene Aliases: 4932409E18; 9430010P06Rik; CLN14; EPM3; KCTD7
UniProt ID: (Human) Q96MP8, (Mouse) Q8BJK1, (Rat) B1WC97
Entrez Gene ID: (Human) 154881, (Mouse) 212919, (Rat) 688993
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support