Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MRRVTLFLNG SPKNGKVVAV YGTLSDLLSV ASSKLGIKAT SVYNGKGGLI DDIALIRDDD VLFVCEGEPF IDPQTDSKPP EGLLGFHTDW LTLNVGGRYF TTTRSTLVNK EPDSMLAHMF KDKGVWGNKQ DHRGAFLIDR SPEYFEPILN YLRHGQLIVN DGINLLGVLE EARFFGIDSL IEHLEVAIKN SQPPEDHSPI SRKEFVRFLL ATPTKSELRC QGLNFSGADL SRLDLRYINF KMANLSRCNL AHANLCCANL ERADLSGSVL DCANLQGVKM LCSNAEGASL KLCNFEDPSG LKANLEGANL KGVDMEGSQM TGINLRVATL KNAKLKNCNL RGATLAGTDL ENCDLSGCDL QEANLRGSNV KGAIFEEMLT PLHMSQSVR
Substrate-specific adapter of a BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex, which mediates the ubiquitination of target proteins, leading to their degradation by the proteasome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BTB/POZ domain-containing protein KCTD9; potassium channel tetramerisation domain containing 9
Gene Aliases: BTBD27; KCTD9
UniProt ID: (Human) Q7L273
Entrez Gene ID: (Human) 54793
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support