Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TQGKLEEKL QELEANPPSD VYLSSRDRQI LDWHFANLEF ANATPLSTLS LKHWDQDDDF EFTGSHLTVR NGYSCVPVAL AEGLDIKLNT AVRQVRYTAS GCEVIAVNTR STSQTFIYKC DAVLCTLPLG VLKQQPPAVQ FVPPLPEWKT SAVQRMGFGN LNKVVLCFDR VFWDPSVNLF GHVGSTTASR GELFLFWNLY KAPILLALVA GEAAGIMENI SDDVIVGRCL AILKGIFGSS AVPQPKETVV SRWRADPWAR GSYSYVAAGS SGNDYDLMAQ PITPGPSIPG APQPIPRLFF AGEHTIRNYP ATVHGALLSG LREAGRIADQ FLGAMYTLPR QATPGVPAQQ SPSM (524-876 aa encoded by B C040194)
LSD1 is a histone demethylase that specifically demethylates 'Lys-4' of histone H3, a specific tag for epigenetic transcriptional activation, thereby acting as a corepressor. LSD1 contains a SWIRM domain, a FAD-binding motif, and an amine oxidase domain. This protein is a component of several histone deacetylase complexes, though it silences genes by functioning as a histone demethylase. It acts by oxidizing the substrate by FAD to generate the corresponding imine that is subsequently hydrolyzed. LSD1 demethylates both mono- and tri-methylted 'Lys-4' of histone H3. This protein may play a role in the repression of neuronal genes. Alone, it is unable to demethylate H3 'Lys-4' on nucleosomes and requires the presence of RCOR1/CoREST to achieve such activity. It may also demethylate 'Lys-9' of histone H3, a specific tag for epigenetic transcriptional repression, thereby leading to derepression of androgen receptor target genes. Mutations affecting this gene can result in cleft palate, psychomotor retardation, and distintive facial features.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [histone H3]-dimethyl-L-lysine(4) FAD-dependent demethylase 1A; amine oxidase (flavin containing) domain 2; BRAF35-HDAC complex protein BHC110; FAD-binding protein BRAF35-HDAC complex, 110 kDa subunit; Flavin-containing amine oxidase domain-containing protein 2; lysine (K)-specific demethylase 1; lysine (K)-specific demethylase 1A; lysine-specific histone demethylase 1; Lysine-specific histone demethylase 1A; RP1-184J9.1
Gene Aliases: 1810043O07Rik; AA408884; AOF2; BHC110; CPRF; D4Ertd478e; KDM1; KDM1A; KIAA0601; LSD1; mKIAA0601
UniProt ID: (Human) O60341, (Mouse) Q6ZQ88
Entrez Gene ID: (Human) 23028, (Mouse) 99982
Molecular Function:
oxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support