Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 8
MA5-31624 has been tested in indirect ELISA.
Flavin-dependent histone demethylases, such as KDM1B, regulate histone lysine methylation, an epigenetic mark that regulates gene expression and chromatin function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: amine oxidase (flavin containing) domain 1; amine oxidase, flavin containing 1; Flavin-containing amine oxidase domain-containing protein 1; lysine (K)-specific demethylase 1B; Lysine-specific histone demethylase 1B; Lysine-specific histone demethylase 2
Gene Aliases: AOF1; bA204B7.3; C6orf193; dJ298J15.2; KDM1B; LSD2
UniProt ID: (Human) Q8NB78
Entrez Gene ID: (Human) 221656
Molecular Function: oxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support