Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Immunogen sequence: HAYSSASKSC SQGSSTPRKQ PRKSPLVPRS LEPPVLELSP GAKAQLEELM MVGDLLEVSL DETQHIWRIL QATHPPSEDR FLHIMEDDSM EEKPLKVKGK DSSEKKRKRK LEKVEQLFGE GKQKSKELKK MDKPRKKKLK LGADKSKELN KLAKKLAKEE ERKKKKEKAA AAKVELVKES TEKKREKKVL DIPSKYDWSG AEESDDENAV CAAQNCQRPC KDKVDWVQCD GGCDEWFHQV CVGVSPEMAE NEDYICINCA KKQGPVSPGP APPPSFIMSY KLPMEDLKET S
UPF3B is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. The protein is one of two functional homologs to yeast Upf3p. This protein binds to the mRNA and remains bound after nuclear export, acting as a nucleocytoplasmic shuttling protein. It forms with Y14 a complex that binds specifically 20 nt upstream of exon-exon junctionsThis gene encodes a protein that is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. The encoded protein is one of two functional homologs to yeast Upf3p. mRNA surveillance detects exported mRNAs with truncated open reading frames and initiates nonsense-mediated mRNA decay (NMD). When translation ends upstream from the last exon-exon junction, this triggers NMD to degrade mRNAs containing premature stop codons. This protein binds to the mRNA and remains bound after nuclear export, acting as a nucleocytoplasmic shuttling protein. It forms with Y14 a complex that binds specifically 20 nt upstream of exon-exon junctions. This gene is located on the long arm of chromosome X. Two splice variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [histone H3]-trimethyl-L-lysine(4) demethylase 5A; Histone demethylase JARID1A; jumonji, AT rich interactive domain 1A (Rbp2 like); Jumonji, AT rich interactive domain 1A (RBP2-like); Jumonji/ARID domain-containing protein 1A; lysine (K)-specific demethylase 5A; Lysine-specific demethylase 5A; RBBP-2; retinoblastoma binding protein 2; Retinoblastoma-binding protein 2
Gene Aliases: AA409370; C76986; JARID1A; KDM5A; RBBP-2; RBBP2; RBP2
UniProt ID: (Human) A8MV76, (Mouse) Q3UXZ9
Entrez Gene ID: (Human) 5927, (Rat) 312678, (Mouse) 214899
Molecular Function: histone modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support