Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Antibody detects endogenous levels of total Keap1.
This gene encodes a protein containing KELCH-1 like domains, as well as a BTB/POZ domain. Kelch-like ECH-associated protein 1 interacts with NF-E2-related factor 2 in a redox-sensitive manner and the dissociation of the proteins in the cytoplasm is followed by transportation of NF-E2-related factor 2 to the nucleus. This interaction results in the expression of the catalytic subunit of gamma-glutamylcysteine synthetase. Two alternatively spliced transcript variants encoding the same isoform have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytosolic inhibitor of Nrf2; INrf2; Kelch-like ECH-associated protein 1; kelch-like family member 19; Kelch-like protein 19; NRF2 cytosolic inhibitor; ring canal protein
Gene Aliases: INRF2; KEAP1; KIAA0132; KLHL19; mKIAA0132
UniProt ID: (Human) Q14145, (Mouse) Q9Z2X8
Entrez Gene ID: (Human) 9817, (Mouse) 50868, (Rat) 117519
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support