Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEEKQILCV GLVVLDVISL VDKYPKEDSE IRCLSQRWQR GGNASNSCTI LSLLGAPCAF MGSMAPGHVA DFVLDDLRRY SVDLRYTVFQ TTGSVPIATV IINEASGSRT ILYYDRSLPD VSATDFEKVD LTQFKWIHIE GRNASEQVKM LQRIDAHNTR QPPEQKIRVS VEVEKPREEL FQLFGYGDVV FVSKDVAKHL GFQSAEEALR GLYGRVRKGA VLVCAWAEEG ADALGPDGKL LHSDAFPPPR VVDTLGAGDT FNASVIFSLS QGRSVQEALR FGCQVAGKKC GLQGFDGIV (1-298 aa encoded by BC006233 )
KHK encodes the gene ketohexokinase that catalyzes conversion of fructose to fructose-1-phosphate. The splice variant presented encodes the highly active form found in liver. renal cortex. and small intestine. while the alternate variant encodes the lowe
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fructokinase; Hepatic fructokinase; Ketohexokinase; ketohexokinase (fructokinase); testicular tissue protein Li 102
Gene Aliases: KETHPRO; KHK
UniProt ID: (Human) P50053, (Mouse) P97328, (Rat) Q02974
Entrez Gene ID: (Human) 3795, (Mouse) 16548, (Rat) 25659
Molecular Function:
carbohydrate kinase
kinase
transferase
metabolite interconversion enzyme
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support